4-(4-Morpholin-4-ylphenyl)-1,3-thiazol-2-amine

Lieferant: Apollo Scientific
Achtung

OR110631-1G
APOSOR110631-1GEA 371 EUR
APOSOR110631-1G
4-(4-Morpholin-4-ylphenyl)-1,3-thiazol-2-amine
4-(4-Morpholin-4-ylphenyl)-1,3-thiazol-2-amine
Formel: C₁₃H₁₅N₃OS
Molecular Weight: 261,343 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17168293
CAS-Nummer: 436152-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR