Ethyl 5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific

OR110633-1G OR110633-10G OR110633-5G
APOSOR110633-1GEA 0 EUR
APOSOR110633-1G APOSOR110633-10G APOSOR110633-5G
Ethyl 5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate
Ethyl 5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate
Formel: C₁₀H₁₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD06662279
CAS-Nummer: 19282-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR