Benzyl 2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR110636-1G
APOSOR110636-1GEA 296 EUR
APOSOR110636-1G
Benzyl 2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate
Benzyl 2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate
Formel: C₁₄H₁₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18074543
CAS-Nummer: 1355171-29-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR