5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbaldehyde

Lieferant: Apollo Scientific

OR110638-1G
APOSOR110638-1GEA 371 EUR
APOSOR110638-1G
5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbaldehyde
5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbaldehyde
Formel: C₈H₈OS
Molecular Weight: 152,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17276640
CAS-Nummer: 956010-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR