Ethyl 5-[(acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific

OR110639-1G
APOSOR110639-1GEA 198 EUR
APOSOR110639-1G
Ethyl 5-[(acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylate
Ethyl 5-[(acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylate
Formel: C₁₃H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD01240570
CAS-Nummer: 100619-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR