4-Hydrazino-5-thien-2-ylthieno[2,3-d]pyrimidine

Lieferant: Apollo Scientific

OR110641-1G
APOSOR110641-1GEA 296 EUR
APOSOR110641-1G
4-Hydrazino-5-thien-2-ylthieno[2,3-d]pyrimidine
4-Hydrazino-5-thien-2-ylthieno[2,3-d]pyrimidine
Formel: C₁₀H₈N₄S₂
Molecular Weight: 248,327 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03444393
CAS-Nummer: 406200-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR