5-[(Acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylic acid
Lieferant: Apollo Scientific
OR110644-1G
APOSOR110644-1GEA
346
EUR
APOSOR110644-1G
5-[(Acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylic acid
5-[(Acetyloxy)methyl]-4-ethyl-3-methyl-1H-pyrrole-2-carboxylic acid
Formel:
C₁₁H₁₅NO₄ Molecular Weight: 225,241 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18804159 CAS-Nummer: 1427460-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...