tert-Butyl 2-{[(1-acetyl-2,3-dihydro-1H-indol-5-yl)sulphonyl]amino}ethylcarbamate
Lieferant: Apollo Scientific
OR110645-1G
APOSOR110645-1GEA
248
EUR
APOSOR110645-1G
tert-Butyl 2-{[(1-acetyl-2,3-dihydro-1H-indol-5-yl)sulphonyl]amino}ethylcarbamate
tert-Butyl 2-{[(1-acetyl-2,3-dihydro-1H-indol-5-yl)sulphonyl]amino}ethylcarbamate
Formel:
C₁₇H₂₅N₃O₅S Molecular Weight: 383,462 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18804160 CAS-Nummer: 1427461-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...