tert-Butyl 2-[(2-bromobenzoyl)amino]ethylcarbamate

Lieferant: Apollo Scientific

OR110647-1G
APOSOR110647-1GEA 248 EUR
APOSOR110647-1G
tert-Butyl 2-[(2-bromobenzoyl)amino]ethylcarbamate
tert-Butyl 2-[(2-bromobenzoyl)amino]ethylcarbamate
Formel: C₁₄H₁₉BrN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18804162
CAS-Nummer: 561030-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR