tert-Butyl 2-[(4-bromobenzoyl)amino]ethylcarbamate

Lieferant: Apollo Scientific

OR110648-1G
APOSOR110648-1GEA 248 EUR
APOSOR110648-1G
tert-Butyl 2-[(4-bromobenzoyl)amino]ethylcarbamate
tert-Butyl 2-[(4-bromobenzoyl)amino]ethylcarbamate
Formel: C₁₄H₁₉BrN₂O₃
Molecular Weight: 343,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18804163
CAS-Nummer: 1008505-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR