N,1-Dimethyl-1H-1,2,3-benzotriazole-5-carboxamide

Lieferant: Apollo Scientific

OR110650-1G
APOSOR110650-1GEA 310 EUR
APOSOR110650-1G
N,1-Dimethyl-1H-1,2,3-benzotriazole-5-carboxamide
N,1-Dimethyl-1H-1,2,3-benzotriazole-5-carboxamide
Formel: C₉H₁₀N₄O
Molecular Weight: 190,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04149706
CAS-Nummer: 722470-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR