4,5,6,7-Tetrahydrobenzo[b]thiophene

Lieferant: Apollo Scientific
Achtung

OR110654-1G
APOSOR110654-1GEA 186 EUR
APOSOR110654-1G
4,5,6,7-Tetrahydrobenzo[b]thiophene
4,5,6,7-Tetrahydrobenzo[b]thiophene
Formel: C₈H₁₀S
Molecular Weight: 138,228 g/mol
Siedepunkt: 52…53 °C/3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD19442759
CAS-Nummer: 13129-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR