Methyl 3-amino-4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)benzoate

Lieferant: Apollo Scientific

OR110655-1G
APOSOR110655-1GEA 371 EUR
APOSOR110655-1G
Methyl 3-amino-4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)benzoate
Methyl 3-amino-4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)benzoate
Formel: C₁₅H₂₃N₃O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD19442760
CAS-Nummer: 1198093-71-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR