Methyl 1-{2-[(tert-butoxycarbonyl)amino]ethyl}-1H-benzimidazole-5-carboxylate

Lieferant: Apollo Scientific

OR110657-1G
APOSOR110657-1GEA 395 EUR
APOSOR110657-1G
Methyl 1-{2-[(tert-butoxycarbonyl)amino]ethyl}-1H-benzimidazole-5-carboxylate
Methyl 1-{2-[(tert-butoxycarbonyl)amino]ethyl}-1H-benzimidazole-5-carboxylate
Formel: C₁₆H₂₁N₃O₄
Molecular Weight: 319,356 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442762
CAS-Nummer: 1427460-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR