1-{2-[(tert-Butoxycarbonyl)amino]ethyl}-1H-benzimidazole-5-carboxylic acid

Lieferant: Apollo Scientific

OR110658-1G
APOSOR110658-1GEA 445 EUR
APOSOR110658-1G
1-{2-[(tert-Butoxycarbonyl)amino]ethyl}-1H-benzimidazole-5-carboxylic acid
1-{2-[(tert-Butoxycarbonyl)amino]ethyl}-1H-benzimidazole-5-carboxylic acid
Formel: C₁₅H₁₉N₃O₄
Molecular Weight: 305,329 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442763
CAS-Nummer: 1427461-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR