4'-Methyl-3-chloropropiophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-1-(p-tolyl)propan-1-one, 3-Chloro-1-(4-methylphenyl)propan-1-one

OR11066-1G
APOSOR11066-1GEA 36.4 EUR
APOSOR11066-1G
4'-Methyl-3-chloropropiophenone
4'-Methyl-3-chloropropiophenone
Formel: C₁₀H₁₁ClO
Molecular Weight: 182,647 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00028195
CAS-Nummer: 22422-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR