1-(Carboxymethyl)cyclopentane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Carboxycyclopent-1-yl)acetic acid, 2-(1-Carboxycyclopent-1-yl)ethanoic acid

OR110660-1G
APOSOR110660-1GEA 194 EUR
APOSOR110660-1G
1-(Carboxymethyl)cyclopentane-1-carboxylic acid
1-(Carboxymethyl)cyclopentane-1-carboxylic acid
Formel: C₈H₁₂O₄
Molecular Weight: 172,178 g/mol
Schmelzpunkt: 155…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00661792
CAS-Nummer: 62953-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR