2-Phenyl-2-azaspiro[4.4]nonane-1,3-dione

Lieferant: Apollo Scientific

OR110662-1G OR110662-5G
APOSOR110662-1GEA 248 EUR
APOSOR110662-1G APOSOR110662-5G
2-Phenyl-2-azaspiro[4.4]nonane-1,3-dione
2-Phenyl-2-azaspiro[4.4]nonane-1,3-dione
Formel: C₁₄H₁₅NO₂
Molecular Weight: 229.274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037419
CAS-Nummer: 856068-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR