N,N-Diethyl-N'-(4-morpholin-4-ylphenyl)thiourea

Lieferant: Apollo Scientific

OR110663-1G
APOSOR110663-1GEA 296 EUR
APOSOR110663-1G
N,N-Diethyl-N'-(4-morpholin-4-ylphenyl)thiourea
N,N-Diethyl-N'-(4-morpholin-4-ylphenyl)thiourea
Formel: C₁₅H₂₃N₃OS
Molecular Weight: 293,428 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037420
CAS-Nummer: 1427460-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR