Ethyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate
Lieferant: Apollo Scientific
OR110671-1G
APOSOR110671-1GEA
371
EUR
APOSOR110671-1G
Ethyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate
Ethyl 2-pyridin-3-yl-1,3-thiazole-4-carboxylate
Formel:
C₁₁H₁₀N₂O₂S Molecular Weight: 234,274 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052302 CAS-Nummer: 39067-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...