N,N-Dimethyl-4-(1H-pyrrol-1-yl)benzenesulphonamide

Lieferant: Apollo Scientific

OR110672-1G
APOSOR110672-1GEA 248 EUR
APOSOR110672-1G
N,N-Dimethyl-4-(1H-pyrrol-1-yl)benzenesulphonamide
N,N-Dimethyl-4-(1H-pyrrol-1-yl)benzenesulphonamide
Formel: C₁₂H₁₄N₂O₂S
Molecular Weight: 250,317 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215314
CAS-Nummer: 338773-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR