N-(Tetrahydrofuran-2-ylmethyl)-1,3-benzothiazole-6-sulphonamide

Lieferant: Apollo Scientific

OR110674-1G
APOSOR110674-1GEA 346 EUR
APOSOR110674-1G
N-(Tetrahydrofuran-2-ylmethyl)-1,3-benzothiazole-6-sulphonamide
N-(Tetrahydrofuran-2-ylmethyl)-1,3-benzothiazole-6-sulphonamide
Formel: C₁₂H₁₄N₂O₃S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18804165
CAS-Nummer: 1427460-47-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR