5-Methyl-N-(tetrahydrofuran-2-ylmethyl)thiophene-2-sulphonamide

Lieferant: Apollo Scientific

OR110675-1G
APOSOR110675-1GEA 296 EUR
APOSOR110675-1G
5-Methyl-N-(tetrahydrofuran-2-ylmethyl)thiophene-2-sulphonamide
5-Methyl-N-(tetrahydrofuran-2-ylmethyl)thiophene-2-sulphonamide
Formel: C₁₀H₁₅NO₃S₂
Molecular Weight: 261,361 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17658281
CAS-Nummer: 1427461-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR