4-(4,5,6,7-Tetrahydro-1-benzothien-2-ylsulphonyl)morpholine

Lieferant: Apollo Scientific

OR110676-1G
APOSOR110676-1GEA 493 EUR
APOSOR110676-1G
4-(4,5,6,7-Tetrahydro-1-benzothien-2-ylsulphonyl)morpholine
4-(4,5,6,7-Tetrahydro-1-benzothien-2-ylsulphonyl)morpholine
Formel: C₁₂H₁₇NO₃S₂
Molecular Weight: 287,398 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21602388
CAS-Nummer: 1427461-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR