4-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]morpholine

Lieferant: Apollo Scientific

OR110677-1G
APOSOR110677-1GEA 395 EUR
APOSOR110677-1G
4-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]morpholine
4-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]morpholine
Formel: C₈H₁₂N₂O₃S₂
Molecular Weight: 248,322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21602389
CAS-Nummer: 1427460-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR