4-(3-Bromophenyl)-2-butanone

Lieferant: Apollo Scientific
Achtung

OR110679-1G
APOSOR110679-1GEA 232 EUR
APOSOR110679-1G
4-(3-Bromophenyl)-2-butanone
4-(3-Bromophenyl)-2-butanone
Formel: C₁₀H₁₁BrO
Molecular Weight: 227,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09991820
CAS-Nummer: 3506-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR