6-(2-Furyl)-2-hydroxy-4-(2-thienyl)nicotinonitrile

Lieferant: Apollo Scientific

OR110681-1G
APOSOR110681-1GEA 125 EUR
APOSOR110681-1G
6-(2-Furyl)-2-hydroxy-4-(2-thienyl)nicotinonitrile
6-(2-Furyl)-2-hydroxy-4-(2-thienyl)nicotinonitrile
Formel: C₁₄H₈N₂O₂S
Molecular Weight: 268,291 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774476
CAS-Nummer: 111121-81-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR