5-Chloro-6-methyl-2-(2-pyridyl)pyrimidin-4-ol

Lieferant: Apollo Scientific

OR110684-1G
APOSOR110684-1GEA 162 EUR
APOSOR110684-1G
5-Chloro-6-methyl-2-(2-pyridyl)pyrimidin-4-ol
5-Chloro-6-methyl-2-(2-pyridyl)pyrimidin-4-ol
Formel: C₁₀H₈ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00113781
CAS-Nummer: 286430-67-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR