Ethyl 5-cyano-1,6-dihydro-6-imino-4-methyl-1-(4-methylphenyl)pyridazine-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Cyano-1,6-dihydro-3-(ethoxycarbonyl)-6-imino-4-methyl-1-(4-methylphenyl)pyridazine, 4-[5-Cyano-3-(ethoxycarbonyl)-6-imino-4-methylpyridazin-1(6H)-yl]toluene
OR110687-10G
OR110687-1G
OR110687-5G
APOSOR110687-10GEA
346
EUR
APOSOR110687-10G
APOSOR110687-1G
APOSOR110687-5G
Ethyl 5-cyano-1,6-dihydro-6-imino-4-methyl-1-(4-methylphenyl)pyridazine-3-carboxylate
Ethyl 5-cyano-1,6-dihydro-6-imino-4-methyl-1-(4-methylphenyl)pyridazine-3-carboxylate
Formel:
C₁₆H₁₆N₄O₂ Schmelzpunkt: 150 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00828950 CAS-Nummer: 120049-79-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...