3',4'-Dichlorovalerophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3,4-Dichlorophenyl)pentan-1-one
OR11069-1G
OR11069-5G
OR11069-10G
APOSOR11069-1GEA
50.9
EUR
APOSOR11069-1G
APOSOR11069-5G
APOSOR11069-10G
3',4'-Dichlorovalerophenone
3',4'-Dichlorovalerophenone
Formel:
C₁₁H₁₂Cl₂O Molecular Weight: 231.118 g/mol Schmelzpunkt: 40…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445511 CAS-Nummer: 68120-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...