4-(Morpholin-4-ylmethyl)benzonitril
Lieferant: Apollo Scientific
Synonyme:
4-(Morpholinomethyl)benzonitrile
OR110690-5G
APOSOR110690-5GEA
134
EUR
APOSOR110690-5G
4-(Morpholin-4-ylmethyl)benzonitril
4-(Morpholin-4-ylmethyl)benzonitril
Formel:
C₁₂H₁₄N₂O Molecular Weight: 202.2500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00454271 CAS-Nummer: 37812-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...