6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 6-methyl-2-oxo-1,2,3,4-tetrahydro-4-thioxopyrimidine-5-carboxylate

OR110691-1G
APOSOR110691-1GEA 198 EUR
APOSOR110691-1G
6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester
6-Methyl-2-oxo-4-thioxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester
Formel: C₈H₁₀N₂O₃S
Molecular Weight: 214.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411556
CAS-Nummer: 13996-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR