4-(2-Oxoimidazolidin-1-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR110698-1G OR110698-5G
APOSOR110698-1GEA 0 EUR
APOSOR110698-1G APOSOR110698-5G
4-(2-Oxoimidazolidin-1-yl)benzoic acid
4-(2-Oxoimidazolidin-1-yl)benzoic acid
Formel: C₁₀H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11505356
CAS-Nummer: 167626-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR