5-Chlorobenzoxazole-2-thiol

Lieferant: Apollo Scientific

Synonyme: 5-Chlorobenzo[d]oxazole-2(3H)-thione

OR110701-1G
APOSOR110701-1GEA 0 EUR
APOSOR110701-1G
5-Chlorobenzoxazole-2-thiol
5-Chlorobenzoxazole-2-thiol
Formel: C₇H₄ClNOS
Molecular Weight: 185,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00175241
CAS-Nummer: 22876-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR