6-Chloro-N~4~-ethylpyrimidine-2,4-diamine

Lieferant: Apollo Scientific
Achtung

OR110705-1G
APOSOR110705-1GEA 58 EUR
APOSOR110705-1G
6-Chloro-N~4~-ethylpyrimidine-2,4-diamine
6-Chloro-N~4~-ethylpyrimidine-2,4-diamine
Formel: C₆H₉ClN₄
Molecular Weight: 172,615 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665927
CAS-Nummer: 6316-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR