4-Chloro-6-pyrrolidin-1-ylpyrimidin-2-ylamine

Lieferant: Apollo Scientific

OR110707-1G
APOSOR110707-1GEA 198 EUR
APOSOR110707-1G
4-Chloro-6-pyrrolidin-1-ylpyrimidin-2-ylamine
4-Chloro-6-pyrrolidin-1-ylpyrimidin-2-ylamine
Formel: C₈H₁₁ClN₄
Molecular Weight: 198,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665930
CAS-Nummer: 263276-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR