8-Brom-6-chlorimidazo[1,2-a]pyridin 95%

Lieferant: Apollo Scientific
Achtung

OR110712-5G
APOSOR110712-5GEA 120 EUR
APOSOR110712-5G
8-Brom-6-chlorimidazo[1,2-a]pyridin 95%
8-Brom-6-chlorimidazo[1,2-a]pyridin
Formel: C₇H₄BrClN₂
MDL: MFCD12828062
CAS-Nummer: 957187-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR