6-Bromobenzo[b]thiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR110713-1G
APOSOR110713-1GEA 261 EUR
APOSOR110713-1G
6-Bromobenzo[b]thiophene-2-carbohydrazide
6-Bromobenzo[b]thiophene-2-carbohydrazide
Formel: C₉H₇BrN₂OS
Molecular Weight: 271,134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11615822
CAS-Nummer: 1171927-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR