2-Ethyl-6-nitrobenzoxazol

Lieferant: Apollo Scientific

OR110720-1G
APOSOR110720-1GEA 0 EUR
APOSOR110720-1G
2-Ethyl-6-nitrobenzoxazol
2-Ethyl-6-nitrobenzoxazole
Formel: C₉H₈N₂O₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 13243-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR