3-Bromo-5-chloroimidazo[1,2-a]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific
Gefahr

OR110724-1G
APOSOR110724-1GEA 328 EUR
APOSOR110724-1G
3-Bromo-5-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
3-Bromo-5-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel: C₈H₄BrClN₂O₂
Molecular Weight: 275,487 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846272
CAS-Nummer: 1227955-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR