4-(2-Pyridinyl)-2-pyrimidinamine
Lieferant: Apollo Scientific
|
Achtung
|
OR110729-1G
APOSOR110729-1GEA
114
EUR
APOSOR110729-1G
4-(2-Pyridinyl)-2-pyrimidinamine
4-(2-Pyridinyl)-2-pyrimidinamine
Formel:
C₉H₈N₄ Molecular Weight: 172,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00138711 CAS-Nummer: 66521-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...