Methyl 3-(3-chloro-4-hydroxyphenyl)acrylate

Lieferant: Apollo Scientific
Achtung

OR110737-1G
APOSOR110737-1GEA 404 EUR
APOSOR110737-1G
Methyl 3-(3-chloro-4-hydroxyphenyl)acrylate
Methyl 3-(3-chloro-4-hydroxyphenyl)acrylate
Formel: C₁₀H₉ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16556319
CAS-Nummer: 1266249-44-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR