N'-(2,5-Dimethoxyphenyl)-N-(2-hydroxyethyl)-N-methylurea
Lieferant: Apollo Scientific
OR110739-1G
APOSOR110739-1GEA
0
EUR
APOSOR110739-1G
N'-(2,5-Dimethoxyphenyl)-N-(2-hydroxyethyl)-N-methylurea
N'-(2,5-Dimethoxyphenyl)-N-(2-hydroxyethyl)-N-methylurea
Formel:
C₁₂H₁₈N₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16619603 CAS-Nummer: 1388191-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...