Ethyl-8-brom-6-methylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: Apollo Scientific
Achtung

OR110748-1G
APOSOR110748-1GEA 128 EUR
APOSOR110748-1G
Ethyl-8-brom-6-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-8-brom-6-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₁BrN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09475852
CAS-Nummer: 847446-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR