3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-8-carboxylic acid, 8-Carboxy-3,4-dihydro-3-oxo-2H-benzo[b][1,4]oxazine, 8-Carboxy-3,4-dihydro-3-oxo-2H-1,4-benzoxazine

OR110751-1G
APOSOR110751-1GEA 144 EUR
APOSOR110751-1G
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid
3,4-Dihydro-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid
Formel: C₉H₇NO₄
Molecular Weight: 193,156 g/mol
Schmelzpunkt: 302…305 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11878407
CAS-Nummer: 208772-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR