3,4-Dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dihydro-4-methyl-3-oxo-2H-benzo[b][1,4]oxazine-8-carboxylic acid, 8-Carboxy-3,4-dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine

OR110753-500MG
APOSOR110753-500MGEA 255 EUR
APOSOR110753-500MG
3,4-Dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid
3,4-Dihydro-4-methyl-3-oxo-2H-1,4-benzoxazine-8-carboxylic acid
Formel: C₁₀H₉NO₄
Molecular Weight: 207,183 g/mol
Schmelzpunkt: 207…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12756239
CAS-Nummer: 141762-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR