5-Methyl-1H-1,2,3-benzotriazol-4-amine
Lieferant: Apollo Scientific
|
Achtung
|
OR110760-1G
APOSOR110760-1GEA
406
EUR
APOSOR110760-1G
5-Methyl-1H-1,2,3-benzotriazol-4-amine
5-Methyl-1H-1,2,3-benzotriazol-4-amine
Formel:
C₇H₈N₄ Molecular Weight: 148,165 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00962616 CAS-Nummer: 137434-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...