2-Methoxyisonicotinsäurehydrazid

Lieferant: Apollo Scientific

OR110761-1G
APOSOR110761-1GEA 0 EUR
APOSOR110761-1G
2-Methoxyisonicotinsäurehydrazid
2-Methoxyisonicotinsäurehydrazid
Formel: C₇H₉N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01747192
CAS-Nummer: 19353-97-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR