2-(2-Chlorophenoxy)acetohydrazide 98%

Lieferant: Apollo Scientific
Achtung

OR11077-5G
APOSOR11077-5GEA 21.9 EUR
APOSOR11077-5G
2-(2-Chlorophenoxy)acetohydrazide 98%
2-(2-Chlorophenoxy)acetohydrazide
Formel: C₈H₉ClN₂O₂
Molecular Weight: 200,63 g/mol
Schmelzpunkt: 111…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082786
CAS-Nummer: 36304-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR