2-(3-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole

Lieferant: Apollo Scientific

OR110771-1G
APOSOR110771-1GEA 0 EUR
APOSOR110771-1G
2-(3-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole
2-(3-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole
Formel: C₉H₆BrClN₂O
Molecular Weight: 273,514 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09935794
CAS-Nummer: 944391-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR